Learn More
Medchemexpress LLC 1-Methylinosine-d3 (N1-Methylinosine-d3) | 2140-73-0 | 99.0% | 285.27 | 10 MG
SDP
Catalog No. 5000437331
Encompass_Preferred
1-Methylinosine-d3 is the deuterium-labeled form of 1-Methylinosine. 1-Methylinosine is a modified nucleotide found at position 37 in tRNA, located 3' to the anticodon of eukaryotic tRNA. Deuterium labeling, as seen in this compound, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • It can be used as a tracer in research applications.
  • It serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-004-37331 Supplier Medchemexpress LLC Supplier No. HY113139S10MG
1-Methylinosine-d3 is the deuterium-labeled form of 1-Methylinosine. 1-Methylinosine is a modified nucleotide found at position 37 in tRNA, located 3' to the anticodon of eukaryotic tRNA. Deuterium labeling, as seen in this compound, has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • It can be used as a tracer in research applications.
  • It serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development.