Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylisatin, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
2058-74-4
C9H7NO2
161.16
MFCD00005812
VCYBVWFTGAZHGH-UHFFFAOYSA-N
n-methylisatin,1-methylisatin,1-methyl-1h-indole-2,3-dione,1-methylindoline-2,3-dione,1h-indole-2,3-dione, 1-methyl,1-methyl-indole-2,3-dione,1 methylisatine,n-methylindol-2,3-dione,1-methyl-2,3-dihydro-1h-indole-2,3-dione,1-methyl-2,3-indolinedione
16358
1-methylindole-2,3-dione
CN1C2=CC=CC=C2C(=O)C1=O
2058-74-4
C9H7NO2
161.16
MFCD00005812
VCYBVWFTGAZHGH-UHFFFAOYSA-N
n-methylisatin,1-methylisatin,1-methyl-1h-indole-2,3-dione,1-methylindoline-2,3-dione,1h-indole-2,3-dione, 1-methyl,1-methyl-indole-2,3-dione,1 methylisatine,n-methylindol-2,3-dione,1-methyl-2,3-dihydro-1h-indole-2,3-dione,1-methyl-2,3-indolinedione
16358
1-methylindole-2,3-dione
CN1C2=CC=CC=C2C(=O)C1=O
Quantity 10g
Color Brown to Orange
Melting Point 129.0°C to 133.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 21, 446
Formula Weight 161.16
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Methylisatin
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 218-164-9