Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI 1-Methylnaphthalene, >96.0% (GC), Reagent Grade, 25 mL

Catalog No. 502781562
Encompass_Preferred

1-Methylnaphthalene is a high-purity laboratory reagent supplied as a clear, colorless to light yellow liquid. With a molecular formula of C11H10, this compound is suitable for various laboratory applications requiring a chemical reagent with a purity of at least 96.0% as determined by gas chromatography.

  • Chemical grade: Reagent
  • Purity: ≥ 96.0% (GC)
  • Molecular weight: 142.20 g/mol
  • Physical state: Liquid
  • Boiling point: 241°C
  • Refractive index: 1.62
  • Density: 1.03 (at 20/20°C)
  • Solubility: Insoluble in water; soluble in ether, alcohol, and benzene

Catalog No. 50-278-1562 Supplier Grainger Supplier No. 19KX08
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylnaphthalene is a high-purity laboratory reagent supplied as a clear, colorless to light yellow liquid. With a molecular formula of C11H10, this compound is suitable for various laboratory applications requiring a chemical reagent with a purity of at least 96.0% as determined by gas chromatography.

  • Chemical grade: Reagent
  • Purity: ≥ 96.0% (GC)
  • Molecular weight: 142.20 g/mol
  • Physical state: Liquid
  • Boiling point: 241°C
  • Refractive index: 1.62
  • Density: 1.03 (at 20/20°C)
  • Solubility: Insoluble in water; soluble in ether, alcohol, and benzene

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.