Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylnaphthalene, Spectrum™ Chemical
SDP

Catalog No. 18621088 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-088 500 g
18-621-087 100 g
18-621-089 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-621-088 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M2116500GMGL
Only null left
Add to Cart
Add to Cart

Synonym: Alpha-Methylnaphthalene, Methyl Naphthalene, Methyl-1-Naphthalene

1-Methylnaphthalene, also known as alpha-methylnaphthalene, is a polycyclic aromatic hydrocarbon with a cetane number of zero. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 90-12-0

Specifications

Boiling Point 244.6°C
Packaging Amber Bottle
Quantity 500 g
Percent Purity 94.5%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.