Learn More
Medchemexpress LLC 1-Methylnicotinamide chloride | 1005-24-9 | 98.9% | 172.61 | 100 MG
SDP
Catalog No. 5000436387
Encompass_Preferred
TRIA-662 (Standard) is the analytical standard of TRIA-662, an endogenous metabolite. This product is intended for research and analytical applications. TRIA-662 (1-Methylnicotinamide chloride) has demonstrated antithrombotic and anti-inflammatory activities. It is commonly utilized in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS, as a reference standard.
  • Analytical standard
  • Endogenous metabolite
  • Demonstrates antithrombotic activities
  • Demonstrates anti-inflammatory activities
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used as a reference standard in HPLC, GC, and MS
Catalog No. 50-004-36387 Supplier Medchemexpress LLC Supplier No. HY113527R100MG
TRIA-662 (Standard) is the analytical standard of TRIA-662, an endogenous metabolite. This product is intended for research and analytical applications. TRIA-662 (1-Methylnicotinamide chloride) has demonstrated antithrombotic and anti-inflammatory activities. It is commonly utilized in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS, as a reference standard.
  • Analytical standard
  • Endogenous metabolite
  • Demonstrates antithrombotic activities
  • Demonstrates anti-inflammatory activities
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used as a reference standard in HPLC, GC, and MS