Learn More
Medchemexpress LLC 1-Methylnicotinamide chloride | 1005-24-9 | 98.9% | 172.61 | 25 MG
SDP
Catalog No. 5000438123
Encompass_Preferred
TRIA-662 (Standard) is the analytical standard for TRIA-662, also known as 1-Methylnicotinamide chloride. This product is classified as an endogenous metabolite and is intended for research and analytical applications. It has demonstrated antithrombotic and anti-inflammatory activities. It is suitable for use in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Analytical standard for TRIA-662.
  • Suitable for research and analytical applications.
  • Exhibits antithrombotic activity.
  • Shows anti-inflammatory activity.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-38123 Supplier Medchemexpress LLC Supplier No. 25MGHY113527R
TRIA-662 (Standard) is the analytical standard for TRIA-662, also known as 1-Methylnicotinamide chloride. This product is classified as an endogenous metabolite and is intended for research and analytical applications. It has demonstrated antithrombotic and anti-inflammatory activities. It is suitable for use in qualitative, quantitative, and methodological research experiments, including techniques such as HPLC, GC, and MS.
  • Analytical standard for TRIA-662.
  • Suitable for research and analytical applications.
  • Exhibits antithrombotic activity.
  • Shows anti-inflammatory activity.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments in HPLC, GC, and MS.