Learn More
Selleck Chemical LLC 1-Methylnicotinamide chloride S3346-1g
SDP
Catalog No. 503109877
Encompass_Preferred
1-Methylnicotinamide (1-MNA 3-Carbamoyl-1-methylpyridin-1-ium Trigonellamide) chloride is an active endogenous metabolite of nicotinamide that exhibits anti-inflammatory and anti-thrombotic activities 1-Methylnicotinamide enhances tumor vasculature formation and markedly increases prostacyclin (PGI2) generation
Catalog No. 50-310-9877 Supplier Selleck Chemical LLC Supplier No. S33461G
1-Methylnicotinamide (1-MNA 3-Carbamoyl-1-methylpyridin-1-ium Trigonellamide) chloride is an active endogenous metabolite of nicotinamide that exhibits anti-inflammatory and anti-thrombotic activities 1-Methylnicotinamide enhances tumor vasculature formation and markedly increases prostacyclin (PGI2) generation