Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-methylnicotinamide-d3 chloride | 1218993-18-0 | 99.3% | 175.63 | C7H6D3ClN2O | 5 MG
SDP

Catalog No. 5000263933
Encompass_Preferred

TRIA-662-d3 is the deuterium-labeled form of 1-methylnicotinamide chloride, an endogenous metabolite reported to have antithrombotic and anti-inflammatory activities. It is provided as a stable isotope-labeled reference standard for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.

  • High purity (99.31%).
  • Molecular formula C7H6D3ClN2O; molecular weight 175.63 g/mol.
  • Suitable as internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Intended use as a tracer in pharmacokinetic and metabolic studies.
  • Storage: solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-63933 Supplier Medchemexpress LLC Supplier No. HY113527S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TRIA-662-d3 is the deuterium-labeled form of 1-methylnicotinamide chloride, an endogenous metabolite reported to have antithrombotic and anti-inflammatory activities. It is provided as a stable isotope-labeled reference standard for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses.

  • High purity (99.31%).
  • Molecular formula C7H6D3ClN2O; molecular weight 175.63 g/mol.
  • Suitable as internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Intended use as a tracer in pharmacokinetic and metabolic studies.
  • Storage: solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.