Learn More
Medchemexpress LLC 1-methylnicotinamide-d3 chloride | 1218993-18-0 | 99.4% | 175.63 | 25 MG
SDP
Catalog No. 5000438342
Encompass_Preferred
TRIA-662-d3 is the deuterium labeled TRIA-662. TRIA-662 (1-Methylnicotinamide chloride) is an endogenous metabolite that exhibits antithrombotic and anti-inflammatory activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-38342 Supplier Medchemexpress LLC Supplier No. 25MGHY113527S
TRIA-662-d3 is the deuterium labeled TRIA-662. TRIA-662 (1-Methylnicotinamide chloride) is an endogenous metabolite that exhibits antithrombotic and anti-inflammatory activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.