Learn More
Medchemexpress LLC 1-methylpiperazine-2,2,3,3,5,5,6,6-d8 | 917358-65-7 | MFCD03428101 | 98.0% | 108.21 g/mol | C5H4D8N2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW4512421MG
Encompass_Preferred
1-Methylpiperazine-2,2,3,3,5,5,6,6-d8 is a deuterium-labeled analog of 1-methylpiperazine used as an isotopic internal standard and synthetic building block in analytical and medicinal chemistry. It is suitable for mass spectrometry-based quantitation and tracer studies, and is characterized by molecular formula C5H4D8N2 (108.21 g/mol) and CAS number 917358-65-7.
  • Isotopic enrichment approximately 98% atom D.
  • Suitable for use as an internal standard in mass spectrometry.
  • Useful as a labeled building block for synthetic chemistry.
  • Stable when stored under recommended conditions for labeled compounds.
  • Supplied in small, accurately weighed quantities for analytical use.
Catalog No. 50-002-80960