Learn More
Cayman Chemical 1-Methylpseudouridine, Purity: ≥98%, CAS: 13860-38-3, 50 mg
SDP
Catalog No. 502913580
Encompass_Preferred
A nucleoside analog; increases the thermodynamic and biological stability of RNA; 1-methylpseudouridine-containing mRNA does not induce IFN-β secretion in A549 cells, indicating low immunogenicity; intramuscular or intradermal administration of luciferase mRNA containing 1-methylpseudouridine via lipid complexes increases luciferase activity in mice; has been incorporated as a modification in mRNA that has been used in the development of mRNA-based vaccines
Catalog No. 50-291-3580 Supplier Cayman Chemical Supplier No. 3540150 MG
A nucleoside analog; increases the thermodynamic and biological stability of RNA; 1-methylpseudouridine-containing mRNA does not induce IFN-β secretion in A549 cells, indicating low immunogenicity; intramuscular or intradermal administration of luciferase mRNA containing 1-methylpseudouridine via lipid complexes increases luciferase activity in mice; has been incorporated as a modification in mRNA that has been used in the development of mRNA-based vaccines