Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrazole-3-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. M2642200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2642200MG 200 mg
M26421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2642200MG Supplier TCI America Supplier No. M2642200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25016-20-0
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00464254
InChI Key YBFIKNNFQIBIQZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
PubChem CID 573176
ChEBI CHEBI:74738
IUPAC Name 1-methyl-1H-pyrazole-3-carboxylic acid
SMILES CN1C=CC(=N1)C(O)=O

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 200 mg
Formula Weight 126.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrazole-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.