Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrene 94.0+%, TCI America™
SDP

Catalog No. M0620100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0620100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0620100MG Supplier TCI America Supplier No. M0620100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2381-21-7
Molecular Formula C17H12
Molecular Weight (g/mol) 216.283
MDL Number MFCD00014329
InChI Key KBSPJIWZDWBDGM-UHFFFAOYSA-N
Synonym pyrene, 1-methyl, 3-methylpyrene, 1-methyl pyrene, pyrene, methyl, methylpyrene, unii-ab8420oxqa, ccris 6066, ab8420oxqa, pyrene, 3-methyl, acmc-1cgme
PubChem CID 16932
IUPAC Name 1-methylpyrene
SMILES CC1=C2C=CC3=CC=CC4=C3C2=C(C=C1)C=C4

Specifications

Melting Point 70°C
Color Yellow
Boiling Point 372°C
Quantity 100 mg
Formula Weight 216.28
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrene
RTECSNumber UR2460000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.