Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyridinium-2-aldoxime Chloride 98.0+%, TCI America™
SDP

Catalog No. M081925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M081925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M081925G Supplier TCI America Supplier No. M081925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical reagent with a minimum purity of 98.0% by HPLC and nonaqueous titration.

Chemical Identifiers

CAS 51-15-0
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.61
MDL Number MFCD00011981
InChI Key HIGSLXSBYYMVKI-UHDJGPCESA-N
Synonym pralidoxime chloride, 1-methylpyridinium-2-aldoxime chloride, 2-pam-cl, 2-hydroxyimino methyl-1-methylpyridin-1-ium chloride, 2-hydroxyimino methyl-1-methylpyridinium chloride, 2-formyl 1-methyl pyridinium oxime chloride
PubChem CID 57428996
IUPAC Name (2E)-1-methyl-2-(nitrosomethylidene)-1,2-dihydropyridine hydrochloride
SMILES Cl.CN1C=CC=C\C1=C/N=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 172.61
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyridinium-2-aldoxime Chloride
RTECSNumber UU4200000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.