Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methylpyrrolidine

Catalog No. 69110100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-110-100ML 100 mL
69-110-500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-110-100ML Supplier Sigma Aldrich Supplier No. 69110100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-methylpyrrolidine is extensively used in the synthesis of pyrrolidine based ionic liquids. some of the other reported applications include:
  • Synthesis of ionic liquid electrolytes for primary li/cfx batteries
  • Preparation of ionic liquid as a reaction medium to carry out sulfuric acid-catalyzed conversion of alkynes to ketones
  • Mcm-47, a highly crystalline ferrierite layered silicate, can be synthesized using a diquaternary ammonium salt prepared from the reaction of 1-methylpyrrolidine with 1,4-dibromobutane

Specifications

CAS 120-94-5
Density 0.800 g/mL (at 20°C (literature))
Boiling Point 76°C to 80°C (lit.)
Molecular Formula C5H11N
Refractive Index n20/D 1.425
Linear Formula C5H11N
MDL Number MFCD00003173
Quantity 100 mL
Molecular Weight (g/mol) 85.15
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.