Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylthio-2-propanone, 98+%

Catalog No. p-7027415
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2414906 5 g
AAB2414914 25 g
AAB2414922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2414906 Supplier Thermo Scientific Chemicals Supplier No. B2414906
Only null left
Add to Cart

CAS: 14109-72-9 | C4H8OS | 104.17 g/mol

1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14109-72-9
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.17
MDL Number MFCD00015325
InChI Key UKFADLGENFFWHR-UHFFFAOYSA-N
Synonym 1-methylthio-2-propanone, methylthio acetone, 2-thia-4-pentanone, unii-s6v0dw7bqa, 1-methylsulfanyl propan-2-one, 2-propanone, 1-methylthio, methylthio-2-propanone, acetonyl methyl sulfide, 1-methylsulfanyl acetone, s6v0dw7bqa
PubChem CID 6430713
IUPAC Name 1-methylsulfanylpropan-2-one
SMILES CSCC(C)=O

Specifications

Density 1.023
Boiling Point 69°C to 70°C (30 mmHg)
Flash Point 43°C (109°F)
Odor Stench
Refractive Index 1.474
Quantity 5 g
UN Number UN1224
Beilstein 1737738
Solubility Information Soluble in alcohol and slightly soluble in water.
Formula Weight 104.17
Percent Purity ≥98%
Chemical Name or Material 1-Methylthio-2-propanone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.