Learn More
Medchemexpress LLC 1-Methyluric acid | 708-79-2 | 98.9% | 182.14 | 1 MG
SDP
Catalog No. 5000453533
Encompass_Preferred
1-Methyluric acid acts on the urinary bladder mucosa and increases the blood glucose, insulin, triglyceride, and cholesterol levels. It is intended for research use only and not sold to patients. This compound is useful for research involving metabolic studies and the effects on various biomarkers, as perfusion of the urinary bladder with its solution has been shown to increase serum total cholesterol, true triglyceride, glucose, and insulin levels in Wistar rats.
  • Intended for research use only
  • Solid appearance
  • White to light brown color
  • Storage as powder at -20°C for 3 years, 4°C for 2 years
  • Storage in solvent at -80°C for 6 months, -20°C for 1 month
  • Increases serum total cholesterol, true triglyceride, glucose, and insulin levels in Wistar rats
Catalog No. 50-004-53533 Supplier Medchemexpress LLC Supplier No. HYW0100311MG
Add to Cart
Add to Cart
1-Methyluric acid acts on the urinary bladder mucosa and increases the blood glucose, insulin, triglyceride, and cholesterol levels. It is intended for research use only and not sold to patients. This compound is useful for research involving metabolic studies and the effects on various biomarkers, as perfusion of the urinary bladder with its solution has been shown to increase serum total cholesterol, true triglyceride, glucose, and insulin levels in Wistar rats.
  • Intended for research use only
  • Solid appearance
  • White to light brown color
  • Storage as powder at -20°C for 3 years, 4°C for 2 years
  • Storage in solvent at -80°C for 6 months, -20°C for 1 month
  • Increases serum total cholesterol, true triglyceride, glucose, and insulin levels in Wistar rats