Learn More
Medchemexpress LLC 1-Methyluric acid-d3 | 1189480-64-5 | 185.16 | 1 MG
SDP
Catalog No. 5000452499
Encompass_Preferred
1-Methyluric acid-d3 is the deuterium-labeled form of 1-Methyluric acid. This compound affects the urinary bladder mucosa and elevates blood glucose, insulin, triglyceride, and cholesterol levels. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-52499 Supplier Medchemexpress LLC Supplier No. HYW010031S11MG
1-Methyluric acid-d3 is the deuterium-labeled form of 1-Methyluric acid. This compound affects the urinary bladder mucosa and elevates blood glucose, insulin, triglyceride, and cholesterol levels. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS