Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Methylxanthine | 6136-37-4 | 166.14 | 1 ML
SDP

Catalog No. 5000447274
Encompass_Preferred

1-Methylxanthine is a caffeine derivative that serves as an essential human urinary metabolite of caffeine and theophylline. It enhances the radiosensitivity of tumor cells and is intended for research and analytical applications.

  • Caffeine derivative and human urinary metabolite
  • Enhances radiosensitivity of tumor cells
  • Molecular formula: C6H6N4O2
  • Molecular weight: 166.14
  • Purity: 98.1% (HPLC)
  • Stable for years as powder, months in solvent

Catalog No. 50-004-47274 Supplier Medchemexpress LLC Supplier No. HYW00844910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Methylxanthine is a caffeine derivative that serves as an essential human urinary metabolite of caffeine and theophylline. It enhances the radiosensitivity of tumor cells and is intended for research and analytical applications.

  • Caffeine derivative and human urinary metabolite
  • Enhances radiosensitivity of tumor cells
  • Molecular formula: C6H6N4O2
  • Molecular weight: 166.14
  • Purity: 98.1% (HPLC)
  • Stable for years as powder, months in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.