Learn More
Medchemexpress LLC 1-Methylxanthine | 6136-37-4 | 166.14 | 50 MG
SDP
Catalog No. 5000447726
Encompass_Preferred
A caffeine derivative, this compound is an essential human urinary metabolite of caffeine and theophylline (1,3-dimethylxanthine). It has been shown to enhance the radiosensitivity of tumor cells. This product is for research use only and not for sale to patients.
  • Caffeine derivative
  • Human urinary metabolite of caffeine and theophylline
  • Enhances the radiosensitivity of tumor cells
  • Exhibits similar activities to other naturally occurring methylxanthines
Catalog No. 50-004-47726 Supplier Medchemexpress LLC Supplier No. HYW00844950MG
Add to Cart
Add to Cart
A caffeine derivative, this compound is an essential human urinary metabolite of caffeine and theophylline (1,3-dimethylxanthine). It has been shown to enhance the radiosensitivity of tumor cells. This product is for research use only and not for sale to patients.
  • Caffeine derivative
  • Human urinary metabolite of caffeine and theophylline
  • Enhances the radiosensitivity of tumor cells
  • Exhibits similar activities to other naturally occurring methylxanthines