Learn More
Arcticwhite LLC Glass Coated 96 Square Deep Well, 1 ML/Well, Conical Bottom, 96 Wells, ANSI/SLAS Standard Base and Footprint, Silicon Dioxide, 200 Nm Thick, -80 Deg C to 80 Deg C, 50 Deep Well Plates per Case
SDP
Catalog No. NC0363855
Encompass
Glass Coated 96 Square Deep Well, 1 ML, Conical Bottom, 96 Wells, ANSI/SLAS Standard Base and Footprint, Silicon Dioxide, 200 Nm Thick, -80 Deg C to 80 Deg C, 50 Deep Well Plates per Case. This 96-well deep well microplate has a glass coating for use with hydrophobic compounds. Chemical resistance - resistant, surface properties similar to glass. Thermal range - -80 deg C to 80 deg C. Extractables - low extractables and low leachables. Oxygen transmission rate - reduced. Supplied by ArcticWhite as 50 deep well plates per case.
This item is not returnable. View return policy
Catalog No. NC0363855 Supplier Arcticwhite LLC Supplier No. AWLS219208
Glass Coated 96 Square Deep Well, 1 ML, Conical Bottom, 96 Wells, ANSI/SLAS Standard Base and Footprint, Silicon Dioxide, 200 Nm Thick, -80 Deg C to 80 Deg C, 50 Deep Well Plates per Case. This 96-well deep well microplate has a glass coating for use with hydrophobic compounds. Chemical resistance - resistant, surface properties similar to glass. Thermal range - -80 deg C to 80 deg C. Extractables - low extractables and low leachables. Oxygen transmission rate - reduced. Supplied by ArcticWhite as 50 deep well plates per case.