Learn More
NEST SCIENTIFIC INC 0.1ml 96 Well PCR Plate, Full Skirt, Clear, H1 notch, 100/cs
Catalog No. NC3589202
Encompass_Preferred
Nest Scienticic PCR plates are made of USP grade VI polypropylene in a touch free, ISO 7 cleanroom. Every well on every plate is vacuum tested for pinhole leaks. Ultra thin walls for ideal heat transfer. Non-cytotoxic, DNase, RNase and DNA free. cGMP compliant (21 CFR part 820/821). Ideal for molecular diagnostics.
Catalog No. NC3589202 Supplier NEST SCIENTIFIC INC Supplier No. 402501
Nest Scienticic PCR plates are made of USP grade VI polypropylene in a touch free, ISO 7 cleanroom. Every well on every plate is vacuum tested for pinhole leaks. Ultra thin walls for ideal heat transfer. Non-cytotoxic, DNase, RNase and DNA free. cGMP compliant (21 CFR part 820/821). Ideal for molecular diagnostics.