Learn More
GINKGO BIOWORKS INC (3P) Ginkgo Bioworks™ Cell Free Protein Synthesis Premium kit (E. coli CFPS Premium): 1 mL
Catalog No. NC3933535
Encompass
Ginkgo's CFPS Premium kit is Ginkgo's premier cell free protein synthesis (CFPS) product, optimized for the highest yields and most challenging proteins. This E. coli transcription/translation system is optimized to produce proteins from a T7 RNA Polymerase promoter. Reactions consist of a bacterial cell lysate and a reagent mix that contain all components required for protein production; the user simply provides a DNA template encoding the protein of interest under control of a T7 promoter. The kit also contains a Solubility Enhancer that can help improve expression and folding of challenging protein targets. Contents: E. coli lysate, Reagent Mix (3.3x), Solubility Enhancer (10x), and control plasmid. The 1 mL kit enables 50 reactions of 20 µL, or reaction volumes can be scaled to your specifications. Storage at -80° C. For research use only.
Catalog No. NC3933535 Supplier GINKGO BIOWORKS INC (3P) Supplier No. CFPSKIT 1ML
Ginkgo's CFPS Premium kit is Ginkgo's premier cell free protein synthesis (CFPS) product, optimized for the highest yields and most challenging proteins. This E. coli transcription/translation system is optimized to produce proteins from a T7 RNA Polymerase promoter. Reactions consist of a bacterial cell lysate and a reagent mix that contain all components required for protein production; the user simply provides a DNA template encoding the protein of interest under control of a T7 promoter. The kit also contains a Solubility Enhancer that can help improve expression and folding of challenging protein targets. Contents: E. coli lysate, Reagent Mix (3.3x), Solubility Enhancer (10x), and control plasmid. The 1 mL kit enables 50 reactions of 20 µL, or reaction volumes can be scaled to your specifications. Storage at -80° C. For research use only.