Learn More
Pacer Global LLC 1 mL Cryo Vials, Externally-Threaded Screw Cap, Stargrip Base, Sterile, 100/PK
SDP
Catalog No. 5000198369
Encompass_Preferred
1 mL Cryo Vials, Externally-Threaded Screw Cap, Stargrip Base, Sterile, 100/PK
Catalog No. 50-001-98369 Supplier Pacer Global LLC Supplier No. MLSCVETS1S
1 mL Cryo Vials, Externally-Threaded Screw Cap, Stargrip Base, Sterile, 100/PK