Learn More
HANNA INSTRUMENTS INC 1 mL Graduated Syringe (6 pcs)
SDP
Catalog No. NC1809400
Encompass
HI740143 is a package of 6 graduated syringes for accurate dosing reagents for chemical test kits and photometers.1mL capacity6 syringes per setIdeal for for accurate reagent dosing for CTK and photometers
Catalog No. NC1809400 Supplier HANNA INSTRUMENTS INC Supplier No. HI740143
HI740143 is a package of 6 graduated syringes for accurate dosing reagents for chemical test kits and photometers.1mL capacity6 syringes per setIdeal for for accurate reagent dosing for CTK and photometers