Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ Advance 96-Well Filter Plates With Ion Exchange Membrane - 1 mL, Mustang S membrane (5/pkg)
Supplier:  Cytiva 8172
Encompass
Versatile Format for Multiple Purification StrategiesHigh performance Supor membrane offers optimal support to retain chromatography sorbents while allowing smooth flow of buffersMustang ion exchange membranes deliver efficient and rapid flow rates that result in shorter processing times and more efficient recovery than resin-based technologies.- Multi-well plates containing Mustang membranes can be used to carry out paralell, high throughput ion exchange pre-fractionation of complex samples.New well design results in faster, more uniform filtration rates across the plate with reduced hold-up volumeNew outlet tip geometry minimizes sample leakage and loss during incubation stepsIntrinsic plate properties prevent target molecules from binding to the plate This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0582695