Learn More
USA Scientific Inc 1 mL serological pipet
Catalog No. 502745351
Encompass
1 mL serological pipet, 0.01 mL graduations, standard tip, sterile, 25/sleeve, 1000/case
Catalog No. 50-274-5351 Supplier USA Scientific Inc Supplier No. 56604107
1 mL serological pipet, 0.01 mL graduations, standard tip, sterile, 25/sleeve, 1000/case