Learn More
Vici Precision Sampling, Inc. 1 ML SYRINGE
Catalog No. NC9601039
Encompass_Preferred
Series A-2 Gas Syringes, A-2 features a push-button valve, which allows sample storage to 250 psi in syringes as small as 25 microliter, type: Standard, for 1 ml sample
Catalog No. NC9601039 Supplier Vici Precision Sampling, Inc. Supplier No. 050033
Series A-2 Gas Syringes, A-2 features a push-button valve, which allows sample storage to 250 psi in syringes as small as 25 microliter, type: Standard, for 1 ml sample