Learn More
Biotage 1 ML TIPS W/ 80UL PROPLUS
SDP
Catalog No. NC1798090
Encompass
Box of 96 PhyTip columns (1 ml volume) containing 80 ul of ProPlus (MabSelect SuRe) affinity resin per column
Catalog No. NC1798090 Supplier Biotage Supplier No. PTR918007
Box of 96 PhyTip columns (1 ml volume) containing 80 ul of ProPlus (MabSelect SuRe) affinity resin per column