Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ1-MSH | 72629-65-3 | 99.6% | 1512.74 | 1 MG
SDP

Catalog No. 5000424626
Encompass_Preferred

γ1-MSH is a potent melanocortin MC3 receptor agonist, exhibiting a Ki of 34 nM for the rat MC3 receptor. This compound demonstrates approximately 40-fold selectivity over MC4, with a Ki of 1318 nM.

  • Melanocortin MC3 receptor agonist
  • Ki of 34 nM for the rat MC3 receptor
  • Displays ~40-fold selectivity over MC4 (Ki=1318 nM)
  • Solid appearance
  • White to off-white color
  • Sequence shortening: YVMGHFRWDRF-NH2
  • Recommended storage for powder: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-24626 Supplier Medchemexpress LLC Supplier No. HYP12141MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ1-MSH is a potent melanocortin MC3 receptor agonist, exhibiting a Ki of 34 nM for the rat MC3 receptor. This compound demonstrates approximately 40-fold selectivity over MC4, with a Ki of 1318 nM.

  • Melanocortin MC3 receptor agonist
  • Ki of 34 nM for the rat MC3 receptor
  • Displays ~40-fold selectivity over MC4 (Ki=1318 nM)
  • Solid appearance
  • White to off-white color
  • Sequence shortening: YVMGHFRWDRF-NH2
  • Recommended storage for powder: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture)
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.