Learn More
Medchemexpress LLC γ1-Msh | 72629-65-3 | 99.6% | 1512.74 | 10 MG
SDP
Catalog No. 5000422296
Encompass_Preferred
γ1-MSH is a melanocortin MC3 receptor agonist with high selectivity over MC4 receptors. This product is provided as a white to off-white solid.
  • Melanocortin MC3 receptor agonist
  • Ki of 34 nM for the rat MC3 receptor
  • Approximately 40-fold selectivity over MC4
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-004-22296 Supplier Medchemexpress LLC Supplier No. HYP121410MG
γ1-MSH is a melanocortin MC3 receptor agonist with high selectivity over MC4 receptors. This product is provided as a white to off-white solid.
  • Melanocortin MC3 receptor agonist
  • Ki of 34 nM for the rat MC3 receptor
  • Approximately 40-fold selectivity over MC4
  • White to off-white solid appearance
  • For research use only