Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ1-MSH | 72629-65-3 | 99.6% | 1512.74 g/mol | 5 MG
SDP

Catalog No. 5000423320
Encompass_Preferred

γ1-MSH is a melanocortin MC3 receptor agonist with a Ki of 34 nM for the rat MC3 receptor. It displays approximately 40-fold selectivity over MC4 (Ki=1318 nM).

  • Molecular weight of 1512.74
  • Chemical formula is C72H97N21O14S
  • CAS number is 72629-65-3
  • Appears as a solid
  • Color is white to off-white
  • Sequence shortening: YVMGHFRWDRF-NH2
  • Ships at room temperature in continental US; may vary elsewhere
  • Store sealed and away from moisture: powder at -80°C for 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-23320 Supplier Medchemexpress LLC Supplier No. HYP12145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ1-MSH is a melanocortin MC3 receptor agonist with a Ki of 34 nM for the rat MC3 receptor. It displays approximately 40-fold selectivity over MC4 (Ki=1318 nM).

  • Molecular weight of 1512.74
  • Chemical formula is C72H97N21O14S
  • CAS number is 72629-65-3
  • Appears as a solid
  • Color is white to off-white
  • Sequence shortening: YVMGHFRWDRF-NH2
  • Ships at room temperature in continental US; may vary elsewhere
  • Store sealed and away from moisture: powder at -80°C for 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.