Learn More
Medchemexpress LLC 1-myristoyl-2-hydroxy-sn-glycero-3-pg sodium | 326495-21-0 | MFCD00674365 | 99.0% | 478.49 g·mol⁻¹ | C20H40NaO9P | 25 MG
SDP
Catalog No. 5000280894
Encompass_Preferred
1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (14:0 Lyso PG) used in membrane research and formulation studies. It is commonly employed as a component for preparing micelles, liposomes, and artificial membranes, and in experiments probing membrane biophysics and drug delivery systems.
  • Lysophospholipid useful for micelle and liposome formation.
  • Suitable for studies of membrane biophysics and interactions.
  • High purity (99.0%) for reproducible experimental results.
  • Solid, white to off-white appearance for easy handling.
  • Soluble in common organic solvents such as ethanol and DMSO.
  • Available in small research pack sizes for laboratory use.
  • Store sealed, away from moisture at -20°C for stability.
Catalog No. 50-002-80894 Supplier Medchemexpress LLC Supplier No. HYW33983825MG
1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (14:0 Lyso PG) used in membrane research and formulation studies. It is commonly employed as a component for preparing micelles, liposomes, and artificial membranes, and in experiments probing membrane biophysics and drug delivery systems.
  • Lysophospholipid useful for micelle and liposome formation.
  • Suitable for studies of membrane biophysics and interactions.
  • High purity (99.0%) for reproducible experimental results.
  • Solid, white to off-white appearance for easy handling.
  • Soluble in common organic solvents such as ethanol and DMSO.
  • Available in small research pack sizes for laboratory use.
  • Store sealed, away from moisture at -20°C for stability.