Learn More
Medchemexpress LLC 1-myristoyl-2-hydroxy-sn-glycero-3-pg sodium | 326495-21-0 | MFCD00674365 | 99.0% | 478.49 g/mol | C20H40NaO9P | 5 MG
SDP
Catalog No. 5000280798
Encompass_Preferred
1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (14:0 Lyso PG) sodium salt used in the preparation of micelles, liposomes, and other artificial membranes for membrane modeling and lipid-based drug carrier research. The compound is supplied as a white to off-white solid with high purity and specified storage recommendations.
  • High purity (99.0%) suitable for biochemical research.
  • Solid, white to off-white appearance for straightforward handling.
  • Available in multiple laboratory pack sizes, including 5 mg.
  • Supports formation of micelles, liposomes, and lipid-based carriers.
  • Recommended storage: -20°C sealed; in solvent, -80°C for long-term stability.
Catalog No. 50-002-80798 Supplier Medchemexpress LLC Supplier No. HYW3398385MG
1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (14:0 Lyso PG) sodium salt used in the preparation of micelles, liposomes, and other artificial membranes for membrane modeling and lipid-based drug carrier research. The compound is supplied as a white to off-white solid with high purity and specified storage recommendations.
  • High purity (99.0%) suitable for biochemical research.
  • Solid, white to off-white appearance for straightforward handling.
  • Available in multiple laboratory pack sizes, including 5 mg.
  • Supports formation of micelles, liposomes, and lipid-based carriers.
  • Recommended storage: -20°C sealed; in solvent, -80°C for long-term stability.