Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-myristoyl-2-hydroxy-sn-glycero-3-pg sodium | 326495-21-0 | MFCD00674365 | 99.0% | 478.49 g/mol | C20H40NaO9P | 50 MG
SDP

Catalog No. 5000282544
Encompass_Preferred

1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid research reagent used to model membrane behavior, study lipid signaling, and prepare liposomes or other lipid assemblies. It is the sodium salt of a 14:0 lysophosphatidylglycerol and is supplied as a high-purity solid for biochemical and biophysical applications.

  • High purity: 99.0%.
  • Used in artificial membrane and liposome studies.
  • Forms micelles and other lipid assemblies in solution.
  • Soluble in common organic solvents for formulation.
  • Storage: sealed at -20°C; in solvent -80°C up to 6 months.
  • Available in small research quantities such as 50 MG.

Catalog No. 50-002-82544 Supplier Medchemexpress LLC Supplier No. HYW33983850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid research reagent used to model membrane behavior, study lipid signaling, and prepare liposomes or other lipid assemblies. It is the sodium salt of a 14:0 lysophosphatidylglycerol and is supplied as a high-purity solid for biochemical and biophysical applications.

  • High purity: 99.0%.
  • Used in artificial membrane and liposome studies.
  • Forms micelles and other lipid assemblies in solution.
  • Soluble in common organic solvents for formulation.
  • Storage: sealed at -20°C; in solvent -80°C up to 6 months.
  • Available in small research quantities such as 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.