Learn More
Medchemexpress LLC 1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium | 326495-21-0 | MFCD16621009 | 99.0% | 478.49 g/mol | C20H40NaO9P | 100 MG
SDP
Catalog No. 5000282027
Encompass_Preferred
1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (lyso-phosphatidylglycerol) containing a myristoyl (14:0) chain at the sn-1 position. Supplied as a white to off-white solid, it is intended for lipid research, liposome preparation, and membrane or biochemical studies requiring a 14:0 lyso-PG species.
  • High purity (99.0%) solid for reproducible results.
  • Suitable for liposome formulation and membrane studies.
  • Sodium salt form improves solubility in aqueous buffers.
  • Storage-stable when sealed at -20°C; solvent storage guidelines provided.
  • Available in multiple pack sizes to support different experimental scales.
Catalog No. 50-002-82027 Supplier Medchemexpress LLC Supplier No. HYW339838100MG
1-Myristoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid (lyso-phosphatidylglycerol) containing a myristoyl (14:0) chain at the sn-1 position. Supplied as a white to off-white solid, it is intended for lipid research, liposome preparation, and membrane or biochemical studies requiring a 14:0 lyso-PG species.
  • High purity (99.0%) solid for reproducible results.
  • Suitable for liposome formulation and membrane studies.
  • Sodium salt form improves solubility in aqueous buffers.
  • Storage-stable when sealed at -20°C; solvent storage guidelines provided.
  • Available in multiple pack sizes to support different experimental scales.