Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-myristoyl-2-hydroxy-sn-glycero-3-phosphoglycerol sodium | 326495-21-0 | MFCD00674365 | 99.0% | 478.49 g/mol | C20H40NaO9P | 10 MG
SDP

Catalog No. 5000282521
Encompass_Preferred

1-Myristoyl-2-hydroxy-sn-glycero-3-phosphoglycerol sodium is the sodium salt of a lyso-phosphatidylglycerol bearing a myristoyl (C14) acyl chain. It is supplied as a research reagent for use as a defined lyso-phospholipid standard or reagent in biochemical assays, membrane studies, and lipidomics.

  • Sodium salt of lyso-phosphatidylglycerol with a C14 acyl chain.
  • High purity: 99.0%.
  • Molecular formula C20H40NaO9P and molecular weight 478.49 g/mol.
  • CAS number 326495-21-0 for unambiguous identification.
  • Available in small milligram packages suitable for analytical work.
  • Intended for biochemical assays, lipidomics, and membrane biology applications.

Catalog No. 50-002-82521 Supplier Medchemexpress LLC Supplier No. HYW33983810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Myristoyl-2-hydroxy-sn-glycero-3-phosphoglycerol sodium is the sodium salt of a lyso-phosphatidylglycerol bearing a myristoyl (C14) acyl chain. It is supplied as a research reagent for use as a defined lyso-phospholipid standard or reagent in biochemical assays, membrane studies, and lipidomics.

  • Sodium salt of lyso-phosphatidylglycerol with a C14 acyl chain.
  • High purity: 99.0%.
  • Molecular formula C20H40NaO9P and molecular weight 478.49 g/mol.
  • CAS number 326495-21-0 for unambiguous identification.
  • Available in small milligram packages suitable for analytical work.
  • Intended for biochemical assays, lipidomics, and membrane biology applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.