Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-myristoyl-2-myristoyl-sn-glycero-3-phosphocholine-d27 | 185906-03-0 | 99.0% | 705.10 | C36H45D27NO8P | 5 MG
SDP

Catalog No. 5000223102
Encompass_Preferred

1-Myristoyl-2-myristoyl-sn-glycero-3-phosphocholine-d27 (DMPC-d27) is a deuterium-labeled dimyristoyl phosphatidylcholine used as a tracer and internal standard for quantitative spectroscopic and mass-spectrometric analyses. It is well suited for NMR, GC-MS, and LC-MS applications, and for membrane and lipid research where isotopic labeling improves detection and quantitation.

  • Purity 99.0%.
  • Chemical formula C36H45D27NO8P.
  • Molecular weight 705.10.
  • Deuterium-labeled (d27) for isotopic tracing.
  • Available in small packaging sizes for analytical use.

Catalog No. 50-002-23102 Supplier Medchemexpress LLC Supplier No. HY146881S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Myristoyl-2-myristoyl-sn-glycero-3-phosphocholine-d27 (DMPC-d27) is a deuterium-labeled dimyristoyl phosphatidylcholine used as a tracer and internal standard for quantitative spectroscopic and mass-spectrometric analyses. It is well suited for NMR, GC-MS, and LC-MS applications, and for membrane and lipid research where isotopic labeling improves detection and quantitation.

  • Purity 99.0%.
  • Chemical formula C36H45D27NO8P.
  • Molecular weight 705.10.
  • Deuterium-labeled (d27) for isotopic tracing.
  • Available in small packaging sizes for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.