Learn More
Medchemexpress LLC 1-myristoyl-sn-glycero-3-phosphatidylcholine-d7 | 95.4% | 474.62 | 1 MG
SDP
Catalog No. 5000436476
Encompass_Preferred
LysoPC(14:0/0:0)-d7 is a deuterium labeled lysophospholipid (LyP). It is a monoglycerophospholipid where a phosphorylcholine moiety occupies a glycerol substitution site. This product exhibits a potent antispasmodic effect and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36476 Supplier Medchemexpress LLC Supplier No. HY113123S11MG
LysoPC(14:0/0:0)-d7 is a deuterium labeled lysophospholipid (LyP). It is a monoglycerophospholipid where a phosphorylcholine moiety occupies a glycerol substitution site. This product exhibits a potent antispasmodic effect and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.