Learn More
Medchemexpress LLC 1-N-Boc-3-hydroxyaze | 25g
SDP
Catalog No. 5000202877
Encompass_Preferred
1-N-Boc-3-hydroxyazetidine is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-N-Boc-3-hydroxyazetidine is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs
Catalog No. 50-002-02877 Supplier Medchemexpress LLC Supplier No. HY4014225G
1-N-Boc-3-hydroxyazetidine is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) 1-N-Boc-3-hydroxyazetidine is also a alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs