Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 1-N-BOC-4-aminopiperidine, 87120-72-7, MFCD01076201, 25g

Catalog No. 500125939
Encompass_Preferred

Molecular Formula C10H20N2O2, Purity 98%, Molecular Weight 200.28, Melting Point 44-48°, Boiling Point 80/0.05mm

Catalog No. 50-012-5939 Supplier Matrix Scientific Supplier No. 01259525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C10H20N2O2, Purity 98%, Molecular Weight 200.28, Melting Point 44-48°, Boiling Point 80/0.05mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.