Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Butyl-3-methylimidazolium dicyanamide, 97%

Catalog No. AAH5917518
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5917518 50 g
AAH5917506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5917518 Supplier Thermo Scientific Chemicals Supplier No. H5917518
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 448245-52-1 | C10H15N5 | 205.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448245-52-1
Molecular Formula C10H15N5
Molecular Weight (g/mol) 205.265
MDL Number MFCD07784457
InChI Key ICIVTHOGIQHZRY-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
PubChem CID 11171745
IUPAC Name 1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide
SMILES CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Color Yellow
Quantity 50 g
Sensitivity Moisture sensitive
Formula Weight 205.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-n-Butyl-3-methylimidazolium dicyanamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.