Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Butyl-3-methylimidazolium hexafluoroantimonate, 99%, Thermo Scientific™

Catalog No. AAH2707406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2707406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2707406 Supplier Thermo Scientific Chemicals Supplier No. H2707406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174645-81-9
Molecular Formula C8H15F6N2Sb
Molecular Weight (g/mol) 374.972
MDL Number MFCD07784453
InChI Key CSHAOLBENQQIBR-UHFFFAOYSA-H
Synonym 1-butyl-3-methylimidazolium hexafluoroantimonate, 1-butyl-3-methylimidazolium; hexafluorostibanuide, bmim sbf6, 1-n-butyl-3-methylimidazolium hexafluoroantimonate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluoroantimonate 1-, 1-butyl-3-methylimidazolium hexafluoroantimonate t
PubChem CID 16688641
IUPAC Name 1-butyl-3-methylimidazol-3-ium;hexafluoroantimony(1-)
SMILES CCCCN1C=C[N+](=C1)C.F[Sb-](F)(F)(F)(F)F

Specifications

Quantity 5 g
Formula Weight 374.97
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium hexafluoroantimonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.