Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Octyl-2-pyrrolidone 98.0+%, TCI America™
SDP

Catalog No. O0325500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0325500ML 500 mL
O032525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0325500ML Supplier TCI America Supplier No. O0325500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2687-94-7
Molecular Formula C12H23NO
Molecular Weight (g/mol) 197.32
MDL Number MFCD00074965
InChI Key WPPOGHDFAVQKLN-UHFFFAOYSA-N
PubChem CID 3033871
IUPAC Name 1-octylpyrrolidin-2-one
SMILES CCCCCCCCN1CCCC1=O

Specifications

Melting Point -23°C
Color Yellow
Boiling Point 172°C
Quantity 500 mL
UN Number 1760
Formula Weight 197.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-n-Octyl-2-pyrrolidone
RTECSNumber UY5896000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.