Learn More
Cayman Chemical 1 N6-Ethenoadenosine 5-monop
SDP
Catalog No. 502985114
Encompass_Preferred
Inhibitor of Cdk1/cyclin B and Cdk/p25 (IC50s 600 and 400 nM respectively)
Catalog No. 50-298-5114 Supplier Cayman Chemical Supplier No. 2122710 UMOL
Inhibitor of Cdk1/cyclin B and Cdk/p25 (IC50s 600 and 400 nM respectively)