Learn More
Medchemexpress LLC 1-(naphthalen-2-yl)ethan-1-one | 93-08-3 | MFCD00004108 | 10g
SDP
Catalog No. 5000265952
Encompass_Preferred
2-Acetonaphthone is a synthetic fragrance material 2-Acetonaphthone increases ROS under UVA/sunlight leading to endoplasmic reticulum stress and decreased mitochondrial membrane potential 2-Acetonaphthone can be used as an adulterant in a variety of cosmetics 2-Acetonaphthone can be used for the study of skin keratinization[1]
Catalog No. 50-002-65952 Supplier Medchemexpress LLC Supplier No. HYY181910G
2-Acetonaphthone is a synthetic fragrance material 2-Acetonaphthone increases ROS under UVA/sunlight leading to endoplasmic reticulum stress and decreased mitochondrial membrane potential 2-Acetonaphthone can be used as an adulterant in a variety of cosmetics 2-Acetonaphthone can be used for the study of skin keratinization[1]