Learn More
Medchemexpress LLC 1-Naphthalenamine, N,N-dimethyl- | 86-56-6 | 99.9% | 10 G
SDP
Catalog No. 5000409868
Encompass_Preferred
N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye that can be used in nitrate reduction tests. It appears as a colorless to light yellow liquid with a density of 1.060±0.06 g/cm3.
  • Aromatic amine.
  • Used in nitrate reduction tests.
  • Colorless to light yellow liquid.
  • Suitable for laboratory centrifugation and storage (implied from general storage info).
  • Used as a fluorescent dye.
  • Applicable in labeling chemistry.
  • Functions as a chemical stain.
  • Used as an analytical chemical.
  • Classified as an analytical dye.
Catalog No. 50-004-09868 Supplier Medchemexpress LLC Supplier No. HYD022210G
N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye that can be used in nitrate reduction tests. It appears as a colorless to light yellow liquid with a density of 1.060±0.06 g/cm3.
  • Aromatic amine.
  • Used in nitrate reduction tests.
  • Colorless to light yellow liquid.
  • Suitable for laboratory centrifugation and storage (implied from general storage info).
  • Used as a fluorescent dye.
  • Applicable in labeling chemistry.
  • Functions as a chemical stain.
  • Used as an analytical chemical.
  • Classified as an analytical dye.