Learn More
Medchemexpress LLC 1-Naphthalenamine, N-phenyl- | 90-30-2 | 99.95% | 219.29 | 25 G
SDP
Catalog No. 5000366660
Encompass_Preferred
N-Phenyl-1-naphthylamine is a dye that fluoresces strongly when bound to the inner phospholipid bilayer of Gram-negative bacteria. It can be used to measure outer membrane permeability. This compound is also a fluorescence probe for odorant-binding proteins (OBP) with a dissociation constant of 1.67 μM. It exhibits an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
  • Fluoresces strongly when bound to the inner phospholipid bilayer of gram-negative bacteria.
  • Can be used to measure outer membrane permeability.
  • Functions as a fluorescence probe for odorant-binding proteins (OBP).
  • Exhibits an excitation wavelength of 337 nM.
  • Exhibits an emission wavelength of 407 nM.
Catalog No. 50-003-66660 Supplier Medchemexpress LLC Supplier No. 25GHYW009756
N-Phenyl-1-naphthylamine is a dye that fluoresces strongly when bound to the inner phospholipid bilayer of Gram-negative bacteria. It can be used to measure outer membrane permeability. This compound is also a fluorescence probe for odorant-binding proteins (OBP) with a dissociation constant of 1.67 μM. It exhibits an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
  • Fluoresces strongly when bound to the inner phospholipid bilayer of gram-negative bacteria.
  • Can be used to measure outer membrane permeability.
  • Functions as a fluorescence probe for odorant-binding proteins (OBP).
  • Exhibits an excitation wavelength of 337 nM.
  • Exhibits an emission wavelength of 407 nM.