Learn More
Medchemexpress LLC 1-Naphthalenamine, N-phenyl- | 90-30-2 | 99.95% | 219.29 | 500 G
SDP
Catalog No. 5000451596
Encompass_Preferred
N-Phenyl-1-naphthylamine is a dye that exhibits strong fluorescence when bound to the inner phospholipid bilayer of Gram-negative bacteria and can be used to measure outer membrane permeability. It also functions as a fluorescence probe for odorant-binding proteins (OBP), with a dissociation constant of 1.67 μM. The compound has an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
  • Fluorescence probe for odorant-binding proteins (OBP)
  • Can measure outer membrane permeability in Gram-negative bacteria
  • Exhibits strong fluorescence when bound to inner phospholipid bilayer
Catalog No. 50-004-51596 Supplier Medchemexpress LLC Supplier No. HYW009756500G
N-Phenyl-1-naphthylamine is a dye that exhibits strong fluorescence when bound to the inner phospholipid bilayer of Gram-negative bacteria and can be used to measure outer membrane permeability. It also functions as a fluorescence probe for odorant-binding proteins (OBP), with a dissociation constant of 1.67 μM. The compound has an excitation wavelength of 337 nM and an emission wavelength of 407 nM.
  • Fluorescence probe for odorant-binding proteins (OBP)
  • Can measure outer membrane permeability in Gram-negative bacteria
  • Exhibits strong fluorescence when bound to inner phospholipid bilayer