Learn More
Medchemexpress LLC HY-18570 500mg Medchemexpress, 1-Naphthaleneacetic acid CAS:86-87-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18570500MG
Encompass_Preferred
Medchemexpress, HY-18570 500mg 1-Naphthaleneacetic acid CAS:86-87-3 1-Naphthaleneacetic acid (1-Naphthylacetic acid), a synthetic auxin, can promote plant growth. 1-Naphthaleneacetic acid is also an inhibitor of PLA2, with an IC50 of 13.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2047